Learn More
Medchemexpress LLC Benzyl 2-bromoethyl ether | 1462-37-9 | 215.09 | 100 G

Supplier: Medchemexpress LLC HYW007388100G
Benzyl 2-bromoethyl ether is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It can also be used in the synthesis of other active compounds.
- Biochemical reagent for life science research
- Used in the synthesis of active compounds
- Liquid appearance
- Colorless to light yellow
- Density: 1.36 g/cm³
- Molecular formula: C9H11BrO
- Shipping at room temperature (continental US)
- Storage (pure form): -20°C for 3 years, 4°C for 2 years
- Storage (in solvent): -80°C for 6 months, -20°C for 1 month
- Boiling point: 258.6°C at 760 mmHg
- Flash point: >113°C
- Soluble in DMSO at ≥ 100 mg/mL
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.