Learn More
Medchemexpress LLC Benzyl 2-bromoethyl ether | 1462-37-9 | 215.09 | 25 G

Supplier: Medchemexpress LLC 25GHYW007388
Benzyl 2-bromoethyl ether is a biochemical reagent used as a biological material or organic compound for life science research. It is also employed in the synthesis of other active compounds and as a laboratory chemical for the manufacture of substances. This product is for research use only and has not been fully validated for medical applications.
- Biochemical reagent
- Biological material
- Organic compound for life science research
- Employed in the synthesis of other active compounds
- Laboratory chemical
- Used for the manufacture of substances
- Purity (GC): 99.1%
- Molecular formula: C9H11BrO
- Appearance: Colorless to light yellow liquid
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.