Learn More
Medchemexpress LLC Benzyl 2-bromoethyl ether | 1462-37-9 | 215.09 | 250 G
SDP
Supplier:  Medchemexpress LLC 250GHYW007388
Encompass_Preferred
Benzyl 2-bromoethyl ether is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It can also be used in the synthesis of other active compounds. This product is a colorless to light yellow liquid.
  • Suitable for life science research.
  • Utilized in the synthesis of active compounds.
  • Stable under recommended storage conditions.
  • Store in a cool, well-ventilated area.
  • Protect from direct sunlight and ignition sources.
  • Pure form storage: -20°C for 3 years, 4°C for 2 years.
  • In solvent storage: -80°C for 6 months, -20°C for 1 month.
Catalog No. 50-004-50044