Learn More
Medchemexpress LLC Benzyl ((2S)-1-(((3-bromo-4,5-dihydroisoxazol-5-yl)methyl)amino)-3-(4-hydroxyphenyl)-1-oxopropan-2-yl) | 744198-19-4 | MFCD12032123 | 98.1% | 476.32 g/mol | C21H22BrN3O5 | 100MG
SDP
Supplier:  Medchemexpress LLC HY123290100MG
Encompass_Preferred
KCC009 is a small-molecule transglutaminase 2 (TG2) inhibitor for research use; it induces p53-independent radiosensitization and is supplied as both solid and DMSO solution forms to support in vitro and in vivo studies.
  • Potent transglutaminase 2 (TG2) inhibitor that modulates radiosensitivity.
  • Induces p53-independent radiosensitization in cell models.
  • Available as solid and as DMSO solution for flexible dosing and formulation.
  • High purity (~98.1%) suitable for research applications.
  • Highly soluble in DMSO (≈250 mg/mL) enabling convenient stock preparation.
  • Characterized by CAS 744198-19-4 and molecular formula C21H22BrN3O5.
Catalog No. 50-002-37415