Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzyl (4-aminobicyclo[2.1.1]hexan-1-yl)carbamate hydrochloride | 1354951-84-0 | MFCD20441712 | 282.77 g·mol⁻¹ | C14H19ClN2O2 | 100 MG
SDP

Supplier:  Medchemexpress LLC HYW036860100MG

Encompass_Preferred

Benzyl (4-aminobicyclo[2.1.1]hexan-1-yl)carbamate hydrochloride is a benzyl carbamate hydrochloride salt used as a synthetic intermediate in medicinal chemistry for the preparation of active compounds. It is supplied as a solid powder and should be stored under recommended low-temperature conditions to maintain stability.

  • Used as a synthetic intermediate in medicinal chemistry.
  • Solid powder appearance.
  • Molecular formula C14H19ClN2O2.
  • Molecular weight 282.77 g·mol⁻¹.
  • Recommended storage: powder -20°C (3 years) or 4°C (2 years); in solution -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-99724


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.