Learn More
Medchemexpress LLC Benzyl (5-hydroxypentyl)carbamate | 87905-98-4 | MFCD01863135 | 98.0% | 237.29 g/mol | C13H19NO3 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW142194100MG
Encompass_Preferred
Benzyl (5-hydroxypentyl)carbamate is a carbamate linker and protecting-group reagent commonly used in organic synthesis, peptide and nucleotide modification, and bioconjugation. It is identified by CAS 87905-98-4, has molecular formula C13H19NO3 (MW 237.29 g/mol), and is offered for research use with typical purity around 98% in small laboratory pack sizes.
  • Carbamate linker for protecting-group strategies
  • Suitable for organic synthesis and bioconjugation
  • Typical purity ~98%
  • Molecular formula C13H19NO3; molecular weight 237.29 g/mol
  • Supplied in small research pack sizes (e.g., 100 MG)
Catalog No. 50-002-83704