Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzyl 6-hydroxy-2,2-dimethyltetrahydro-3aH-cyclopenta[d]dioxol-4-ylcarbamate | 274693-53-7 | MFCD07787498 | 98.5% | C16H21NO5 | 100 G
SDP

Supplier:  Medchemexpress LLC HY76819100G

Encompass_Preferred

Benzyl 6-hydroxy-2,2-dimethyltetrahydro-3aH-cyclopenta[d]dioxol-4-ylcarbamate is a chemical compound with the molecular formula C16H21NO5 and a molecular weight of 307.35. Identified by CAS number 274693-53-7 and MFCD07787498, this product is supplied as a white to off-white solid with a purity of 98.47% by HPLC. It is suitable for various research applications.

  • High purity of 98.47% (HPLC)
  • Supplied as a white to off-white solid
  • Stable as powder for up to 3 years at -20°C or 2 years at 4°C
  • When in solvent, stable for 6 months at -80°C or 1 month at -20°C

Catalog No. 50-004-34084


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.