Learn More
Medchemexpress LLC Benzyl acetate | 140-11-4 | 1 ML

Supplier: Medchemexpress LLC HYN712410MM/1ML
Benzyl acetate is an aromatic ester characterized by its jasmine scent. Found naturally in plants, this compound is widely utilized as a base fragrance ingredient in perfumes, cosmetics, and soap fragrances, and is a significant variety in the synthetic fragrance industry.
- Aromatic ester with a jasmine scent
- Found in plants
- Used as a base fragrance ingredient in perfumes, cosmetics, and soap fragrances
- Significant variety in the synthetic fragrance industry
- Purity of 99.94%
- Molecular weight of 150.18
- Chemical formula C9H10O2
- Appears as a liquid
- Colorless to light yellow
- Soluble in DMSO at 125 mg/mL
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.