Learn More
Medchemexpress LLC Benzyl acetate | 140-11-4 | 25 G

Supplier: Medchemexpress LLC 25GHYN7124
Benzyl acetate is an aromatic ester with a jasmine scent, commonly used as a base fragrance ingredient in perfumes, cosmetics, and soap fragrances. It is also one of the largest varieties in the synthetic fragrance industry and is identified for laboratory chemical and substance manufacturing uses. This product is for research use only.
- Molecular formula: C9H10O2
- Molecular weight: 150.18
- Appearance: Colorless to light yellow liquid
- Purity: 99.94% (GC)
- Melting/freezing point: -51 °C
- Boiling point: 213.5 °C at 760 mmHg
- Flash point: 102 °C
- Stable under recommended storage conditions
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.