Learn More
Medchemexpress LLC Benzyl acetate | 140-11-4 | 500 G

Supplier: Medchemexpress LLC HYN7124500G
Benzyl acetate is an aromatic ester with a jasmine scent. It is found in plants and is commonly used as a base fragrance ingredient in perfumes, cosmetics, and soap fragrances. It is also one of the largest varieties in the synthetic fragrance industry.
- Molecular weight: 150.18
- Chemical formula: C9H10O2
- Appearance: Colorless to light yellow liquid
- Purity: 98.0%
- Storage: Store at room temperature for 3 years; in solvent at -80°C for 2 years or -20°C for 1 year
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.