Learn More
Medchemexpress LLC Benzyl acetate | 140-11-4 | MFCD00008712 | 1 KG
SDP
Supplier:  Medchemexpress LLC HYN71241KG
Encompass_Preferred
Benzyl acetate is an aromatic ester with a jasmine scent. It is found in plants and is commonly used as a base fragrance ingredient in perfumes, cosmetics, and soap fragrances. It is also one of the largest varieties in the synthetic fragrance industry.
  • Aromatic ester with a jasmine scent
  • Found in plants
  • Commonly used as a base fragrance ingredient in perfumes, cosmetics, and soap fragrances
  • One of the largest varieties in the synthetic fragrance industry
Catalog No. 50-004-21101