Learn More
Medchemexpress LLC Benzyl acetate | 140-11-4 | MFCD00008712 | 100 G

Supplier: Medchemexpress LLC HYN7124100G
Benzyl acetate is an aromatic ester found in plants, characterized by its jasmine scent. It serves as a primary fragrance ingredient in perfumes, cosmetics, and soap fragrances, and is a significant compound in the synthetic fragrance industry.
- Aromatic ester with a jasmine scent
- Naturally found in plants
- Commonly used as a base fragrance ingredient
- Significant compound in the synthetic fragrance industry
- Molecular weight: 150.18
- Chemical formula: C9H10O2
- Appears as a colorless to light yellow liquid
- Purity: 99.94%
- For research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.