Learn More
Medchemexpress LLC Benzyl alcohol | 100-51-6 | 99.99% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB089210MM/1ML
Encompass_Preferred
Benzyl alcohol is an aromatic alcohol, presenting as a colorless liquid with a mild aromatic odor. It functions as an inhibitor of the P450 enzyme. Research demonstrates its ability to reduce the inflammatory response of liver injury in mice by mediating Toll-Like Receptor 4. It also regulates cyclic AMP synthesis in MDCK cells by increasing membrane fluidity and can inhibit hepatotoxicity by inhibiting cytochrome P450 enzyme activity.
  • Acts as an inhibitor of the P450 enzyme.
  • Reduces the inflammatory response of liver injury in mice by mediating Toll-Like Receptor 4.
  • Regulates cyclic AMP synthesis in MDCK cells by increasing membrane fluidity.
  • Inhibits hepatotoxicity by inhibiting cytochrome P450 enzyme activity.
  • Attenuates the increase in ALT activities and reduces areas of necrosis in APAP-induced liver injury models.
  • Decreases inflammasome activation in a TLR4-dependent manner.
Catalog No. 50-004-09310