Learn More
Medchemexpress LLC Benzyl alcohol | 100-51-6 | 99.99% | 10 G
SDP
Supplier:  Medchemexpress LLC HYB089210G
Encompass_Preferred
Benzyl alcohol is a colorless aromatic alcohol with a mild odor. It functions as an inhibitor of the P450 enzyme and has been shown to reduce inflammatory responses in liver injury models by mediating Toll-Like Receptor 4. This chemical is suitable for various research applications.
  • Inhibits P450 enzyme activity
  • Reduces inflammatory response in liver injury models
  • Regulates cyclic AMP synthesis in MDCK cells
  • Features a purity of 99.99%
  • Colorless to light yellow liquid appearance
Catalog No. 50-004-07677