Learn More
Medchemexpress LLC Benzyl alcohol | 100-51-6 | 99.99% | 5 G
SDP
Supplier:  Medchemexpress LLC HYB08925G
Encompass_Preferred
Benzyl alcohol is an aromatic alcohol, a colorless liquid with a mild aromatic odor. It is known to inhibit the P450 enzyme and has demonstrated the ability to mediate Toll-Like Receptor 4, which can reduce the inflammatory response of liver injury in mice.
  • Aromatic alcohol
  • Colorless liquid with a mild aromatic odor
  • Acts as an inhibitor of the P450 enzyme
  • Shown to mediate Toll-Like Receptor 4
  • Reduces inflammatory response of liver injury in mice
Catalog No. 50-004-09221