Learn More
Medchemexpress LLC Benzyl alcohol-13C6 | 201740-95-6 | 98.0% | 114.09 g/mol | C7H8O (ring-13C6) | 10 MG
SDP
Supplier:  Medchemexpress LLC HYB0892S310MG
Encompass_Preferred
Benzyl alcohol-13C6 is a ring-13C6 isotopically labeled benzyl alcohol used as an internal standard, tracer, or reference material in analytical chemistry and mass spectrometry. It is supplied in small, high-purity quantities suitable for MS calibration and isotopic tracing studies.
  • Isotopically labeled with six 13C atoms in the phenyl ring.
  • High purity: 98.0%.
  • Suitable as an internal standard for mass spectrometry and tracer studies.
  • Available in small quantities for analytical use (e.g., 10 mg).
  • CAS number 201740-95-6 for precise identification.
Catalog No. 50-002-98134