Learn More
Medchemexpress LLC Benzyl alcohol-13C6 | 201740-95-6 | 98% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB0892S31MG
Encompass_Preferred
Benzyl alcohol-13C6 is the 13C labeled Benzyl alcohol. It is for research use only. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • 13C labeled compound
  • Can be used as a tracer and internal standard for quantitative analysis
  • Suitable for NMR, GC-MS, or LC-MS applications
  • Utilizes stable heavy isotopes for drug development quantitation
  • Has potential to affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-09074