Learn More
Medchemexpress LLC Benzyl alcohol-13C6 | 201740-95-6 | 98% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB0892S35MG
Encompass_Preferred
Benzyl alcohol-13C6 is a 13C labeled form of benzyl alcohol designed for research use. This compound acts as a tracer and internal standard for precise quantitative analysis in drug development processes.
  • Used as a tracer for quantitative analysis
  • Functions as an internal standard for quantitative analysis
  • Compatible with NMR, GC-MS, or LC-MS analytical techniques
  • Incorporates stable heavy isotopes into drug molecules
  • Supports quantitation during drug development
Catalog No. 50-004-09075