Learn More
Medchemexpress LLC Benzyl alcohol-13C6 (Benzenemethanol-13C6) | 201740-95-6 | 98% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYB0892S3
Encompass_Preferred
Benzyl alcohol-13C6 is the 13C labeled Benzyl alcohol. It can be used as a tracer or as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • For research use only
Catalog No. 50-004-50805