Learn More
Selleck Chemical LLC Benzyl alcohol S4600-100mg
SDP
Supplier:  Selleck Chemical LLC S4600100MG
Encompass_Preferred
Benzyl alcohol (Phenylmethanol Benzenemethanol Phenylcarbinol Benzoyl alcohol) is used as a local anesthetic Also it is used in the manufacture of other benzyl compounds as a pharmaceutic aid and in perfumery and flavoring
Catalog No. 50-310-8289