Learn More
Medchemexpress LLC Benzyl alcohol-d5 | 68661-10-9 | 99.05% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYB0892S2
Encompass_Preferred
Benzyl alcohol-d5 is the deuterium labeled Benzyl alcohol. It is an aromatic alcohol, presenting as a colorless liquid with a mild, pleasant aromatic odor. This compound is intended for research use only.
  • Can be used as a tracer.
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes are incorporated into drug molecules as tracers for quantitation during drug development.
  • Deuteration can affect pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-50832