Learn More
Medchemexpress LLC Benzyl alcohol-d7 | 71258-23-6 | 99.6% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYB0892S1
Encompass_Preferred
Benzyl alcohol-d7 is the deuterium-labeled Benzyl alcohol, an aromatic alcohol that appears as a colorless liquid with a mild, pleasant aromatic odor. It is primarily identified for use as laboratory chemicals and in the manufacture of substances. This compound is stable under recommended storage conditions and is intended for research use only.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Incorporates stable heavy isotopes as tracers for quantitation in drug development.
  • Deuteration has potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Intended for research use only.
  • Handle by experienced personnel in appropriately equipped and authorized facilities.
Catalog No. 50-004-50725