Learn More
Medchemexpress LLC Benzyl alcohol (Standard) | 100-51-6 | MFCD00004599 | 98.2% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB0892R50MG
Encompass_Preferred
Benzyl alcohol (Standard) is the analytical standard of Benzyl alcohol. This product is intended for research and analytical applications. It is an aromatic alcohol, a colorless liquid with a mild aromatic odor, supplied as a reference standard assay.
  • Analytical standard for research and analytical applications
  • Commonly used in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS applications
  • Colorless liquid with a mild aromatic odor
Catalog No. 50-004-09232