Learn More
TARGETMOL CHEMICALS INC BENZYL-ALPHA-GALNAC 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T1228225MG
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Benzyl-(alpha)-GalNAc (O-glycosylation-IN-1) is a potent inhibitor of O-glycosylation and is used to reduce mucin on cell surfaces. purity: 96%
Catalog No. 50-258-1471