Learn More
Medchemexpress LLC Benzyl benzoate-d12 | 352431-26-6 | 99.85% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB0935S11MG
Encompass_Preferred
Benzyl benzoate-d12 is a deuterium labeled form of Benzyl benzoate. It is used as a fragrance ingredient in cosmetic products and for research into Scabies and Demodex-associated inflammatory skin conditions. This compound serves as a tracer and internal standard in quantitative analysis.
  • Deuterium labeled Benzyl benzoate
  • Functions as a tracer in research
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotope for drug development studies
  • Potential to affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-09092