Learn More
Medchemexpress LLC Benzyl benzoate-d5 | 347840-01-1 | 99.8% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB0935S100MG
Encompass_Preferred
Benzyl benzoate-d5 is the deuterium labeled Benzyl benzoate. Benzyl benzoate (Benzoic acid benzyl ester) is a fragrance ingredient in cosmetic products. It can be used for the research of Scabies and Demodex-associated inflammatory skin conditions. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-08286