Learn More
Medchemexpress LLC Benzyl benzoate-d5 | 347840-01-1 | 99.81% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB0935S5MG
Encompass_Preferred
Benzyl benzoate-d5 is a deuterium-labeled version of Benzyl benzoate (Benzoic acid benzyl ester). It serves as a fragrance ingredient in cosmetic products and is also used in the research of Scabies and Demodex-associated inflammatory skin conditions. This compound can be utilized as a tracer or an internal standard for quantitative analysis, and deuterium substitution can impact the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-09091