Learn More
Medchemexpress LLC Benzyl butyl phthalate | 85-68-7 | 97.0% | 312.36 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYW01133810MM/1ML
Encompass_Preferred
Benzyl butyl phthalate is a member of phthalic acid esters (PAEs). It can trigger the migration and invasion of hemangioma (HA) cells by upregulating Zeb1. Additionally, it activates the aryl hydrocarbon receptor (AhR) in breast cancer cells, stimulating SPHK1/S1P/S1PR3 signaling and enhancing the formation of metastasis-initiating breast cancer stem cells (BCSCs).
  • Can trigger migration and invasion of hemangioma (HA) cells via upregulation of Zeb1.
  • Activates aryl hydrocarbon receptor (AhR) in breast cancer cells.
  • Stimulates SPHK1/S1P/S1PR3 signaling.
  • Enhances the formation of metastasis-initiating breast cancer stem cells (BCSCs).
Catalog No. 50-004-55009