Learn More
Medchemexpress LLC Benzyl butyl phthalate | 85-68-7 | 97.0% | 312.36 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW011338S10MG
Encompass_Preferred
Benzyl butyl phthalate (BBP) is a phthalic acid ester (PAE) that can trigger the migration and invasion of hemangioma (HA) cells by upregulating Zeb1. It also activates aryl hydrocarbon receptor (AhR) in breast cancer cells, stimulating SPHK1/S1P/S1PR3 signaling and enhancing the formation of metastasis-initiating breast cancer stem cells (BCSCs). BBP is commonly added during the manufacturing of plastics to increase flexibility and elasticity.
  • Can trigger the migration and invasion of hemangioma (HA) cells via upregulation of Zeb1.
  • Activates aryl hydrocarbon receptor (AhR) in breast cancer cells.
  • Stimulates SPHK1/S1P/S1PR3 signaling.
  • Enhances the formation of metastasis-initiating breast cancer stem cells (BCSCs).
  • Induces histone modifications in S1PR3 in side population (SP) cells.
  • Commonly added during plastic manufacturing to increase flexibility and elasticity.
  • For research use only.
Catalog No. 50-004-55094