Learn More
Selleck Chemical LLC Benzyl cinnamate S5167-25mg
SDP
Supplier:  Selleck Chemical LLC S516725MG
Encompass_Preferred
Benzyl cinnamate (Cinnamein Benzylcinnamoate Benzyl 3-phenylpropenoate Cinnamic acid benzyl ester) a flavouring agent isolated from various plant species is used in heavy oriental perfumes and as a fixative
Catalog No. 50-311-1991