Learn More
Medchemexpress LLC Benzyl cinnamate | 103-41-3 | 1 ML

Supplier: Medchemexpress LLC HYN709010MM/1ML
Benzyl cinnamate occurs naturally in Balsam of Peru and Tolu balsam, Sumatra and Penang benzoin, and is the main constituent of copaiba balsam. It is utilized in heavy oriental perfumes and as a fixative. This compound exhibits antibacterial activity against S. aureus and S. epidermidis (MIC = 537.81 μM).
- Antibacterial activity against S. aureus and S. epidermidis
- Used in heavy oriental perfumes
- Acts as a fixative
- Molecular weight: 238.29
- Molecular formula: C16H14O2
- Appearance: Off-white to light yellow solid (<34°C) or liquid (>37°C)
- Soluble in DMSO (100 mg/mL)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.