Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzyl D-serinate hydrochloride | 151651-44-4 | MFCD00191017 | 99.4% | 231.68 | C10H14ClNO3 | 100 G
SDP

Supplier:  Medchemexpress LLC HYW035886100G

Encompass_Preferred

Benzyl D-serinate hydrochloride is a benzyl-protected D-serine derivative used in research as a building block or intermediate. It is supplied as a white to off-white solid suitable for synthetic and biological studies involving D-serine derivatives.

  • White to off-white solid.
  • Chemical formula C10H14ClNO3.
  • Molecular weight 231.68.
  • Purity 99.44%.
  • Available in multiple pack sizes including 5 g, 10 g, 25 g, and 100 g.
  • Storage: 4°C, sealed; in solvent, -80°C for 6 months and -20°C for 1 month.

Catalog No. 50-002-99371


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.