Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzyl D-serinate hydrochloride | 151651-44-4 | MFCD00191017 | 99.4% | 231.68 g/mol | C10H14ClNO3 | 25 G
SDP

Supplier:  Medchemexpress LLC 25GHYW035886

Encompass_Preferred

Benzyl D-serinate hydrochloride is the benzyl ester hydrochloride salt of D-serine, an amino-acid derivative used as a research reagent in biochemical and medicinal chemistry applications. It is supplied as a solid with high reported purity and is used as a synthetic building block, analytical standard, and for studies involving serine-related biochemistry.

  • High reported purity (99.44%).
  • Molecular weight 231.68 g/mol.
  • Chemical formula C10H14ClNO3.
  • Solid form suitable for laboratory storage and handling.
  • Available in gram-scale quantities for research use.
  • Useful as a D-serine derivative for synthesis and biochemical studies.

Catalog No. 50-003-04021


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.