Learn More
Medchemexpress LLC Benzyl DC-81 | 127810-79-1 | C20H20N2O3 | 1 ML

Supplier: Medchemexpress LLC HY15346910MM/1ML
Benzyl DC-81 (Compound 6a) is an anticancer agent that exhibits antiproliferative activity against A375 and MCF-7 cells. It has shown cytotoxicity against CH1 human ovarian cells with an IC50 of 1 μM and against L1210 Leukemia cells with an IC50 of 3 μM. This product is for research use only and has not been fully validated for medical applications.
- Anticancer agent
- Exhibits antiproliferative activity against A375 and MCF-7 cells
- Target: ADC payload
- Antibody-drug conjugate/ADC related
- Cytotoxicity against CH1 human ovarian cells with an IC50 of 1 μM
- Cytotoxicity against L1210 leukemia cells with an IC50 of 3 μM
- Consistent with structure by 1H NMR and LCMS
- Purity of 95.80% by LCMS
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.