Learn More
Medchemexpress LLC Benzyl DC-81 | 127810-79-1 | C20H20N2O3 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY153469100MG
Encompass_Preferred
Benzyl DC-81 is an anticancer agent that exhibits antiproliferative activity against A375 and MCF-7 cells. It shows cytotoxicity against CH1 human ovarian cells with an IC50 of 1 μM and against L1210 Leukemia cells with an IC50 of 3 μM. This product is a solid, light yellow to yellow in color, and is soluble in DMSO at 300 mg/mL.
  • Anticancer agent
  • Antiproliferative activity against A375 and MCF-7 cells
  • Cytotoxicity against CH1 human ovarian cells (IC50 = 1 μM)
  • Cytotoxicity against L1210 Leukemia cells (IC50 = 3 μM)
  • Solid, light yellow to yellow appearance
  • Soluble in DMSO at 300 mg/mL
Catalog No. 50-003-72915