Learn More
Medchemexpress LLC Benzyl DC-81 | 127810-79-1 | C20H20N2O3 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY15346950MG
Encompass_Preferred
Benzyl DC-81 (Compound 6a) is an anticancer agent that demonstrates antiproliferative activity against A375 and MCF-7 cells. It has shown cytotoxicity against CH1 human ovarian cells with an IC50 of 1 μM and against L1210 Leukemia cells with an IC50 of 3 μM. This product appears as a light yellow to yellow solid and is soluble in DMSO at a concentration of 300 mg/mL. It is intended for research use only.
  • Anticancer agent
  • Exhibits antiproliferative activity against A375 and MCF-7 cells
  • Cytotoxicity against CH1 human ovarian cells (IC50 = 1 μM)
  • Cytotoxicity against L1210 Leukemia cells (IC50 = 3 μM)
  • Related to ADC payload and cancer targeted therapy
Catalog No. 50-003-72758