Learn More
Medchemexpress LLC Benzyl DC-81 (compound 6a) | 127810-79-1 | C20H20N2O3 | 25 MG

Supplier: Medchemexpress LLC 25MGHY153469
Benzyl DC-81 (compound 6a) is an anticancer agent that exhibits antiproliferative activity against A375 and MCF-7 cells. It is categorized as an ADC Payload, falling under the Antibody-drug Conjugate/ADC Related pathway.
- Exhibits antiproliferative activity against A375 and MCF-7 cells
- Molecular weight is 336.38
- Targeted as an ADC payload
- Appearance as a light yellow to yellow solid
- Soluble in DMSO at 300 mg/mL (891.85 mM)
- Powder storage: -20°C for 3 years
- In solvent storage: -80°C for 6 months, -20°C for 1 month
- Purity (LCMS): 95.80%
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.