Learn More
Medchemexpress LLC Benzyl-α-GalNAc | 3554-93-6 | 99.9% | 10 MG
SDP
Supplier:  Medchemexpress LLC HY12938910MG
Encompass_Preferred
Benzyl-α-GalNAc is a potent O-glycosylation inhibitor. It effectively inhibits the proliferation and activation of LX-2 cells and suppresses the expression of collagen I/III, indicating its potential for investigation in liver fibrosis. Additionally, it significantly enhances the anti-tumor activity of 5-Fluorouracil (e.g., in pancreatic cancer) by inhibiting O-glycosylation.
  • Potent O-glycosylation inhibitor
  • Inhibits proliferation and activation of LX-2 cells
  • Suppresses expression of collagen I/III
  • Potential for investigation in liver fibrosis
  • Enhances anti-tumor activity of 5-Fluorouracil
  • Useful in pancreatic cancer research
Catalog No. 50-003-39712