Learn More
Medchemexpress LLC Benzyl L-leucyl-L-phenylalaninate TFA | 70637-28-4 | C24H29F3N2O5 | 500 MG

Supplier: Medchemexpress LLC HY79862500MG
Benzyl L-leucyl-L-phenylalaninate TFA is a peptide. This compound is specifically for research applications and is not for sale to patients, ensuring its use strictly within scientific studies.
- Classified as a peptide
- Molecular weight: 482.49
- Appears as a solid, white to off-white substance
- Store at 4°C, sealed, and away from moisture
- When in solvent, store at -80°C for 6 months or -20°C for 1 month, sealed and away from moisture
- Soluble in DMSO for in vitro experiments
- Multiple dissolution protocols available for in vivo studies
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.