Learn More
Medchemexpress LLC Benzyl methyl(2-(methylamino)ethyl)carbamate | 148088-78-2 | 98.8% | 222.29 | 1 G
SDP
Supplier:  Medchemexpress LLC HY448161G
Encompass_Preferred
Benzyl methyl(2-(methylamino)ethyl)carbamate is a drug intermediate used for the synthesis of various active compounds.
  • White to light yellow solid
  • Purity confirmed by LCMS
  • Structure consistent with 1H NMR and LCMS
Catalog No. 50-004-01660