Learn More
Medchemexpress LLC Benzyl N-[(2S)-1-[(4-methyl-2-oxochromen-7-yl)amino]-1-oxo-6-(propanoylamino)hexan-2-yl]carbam | 787549-26-2 | MFCD08705325 | 99.6% | 493.55 | C27H31N3O6 | 5 MG

Supplier: Medchemexpress LLC HY1414275MG
MOCPAC is a research-grade small molecule substrate selective for histone deacetylase 1 (HDAC1), intended for use in biochemical and enzymatic assays that require HDAC1-specific activity measurement. The compound is supplied as a solid suitable for formulation and assay preparation.
- HDAC1-specific substrate for biochemical assays.
- Purity 99.6%.
- Molecular weight 493.55.
- Chemical formula C27H31N3O6.
- Appearance white to off-white solid.
- CAS number 787549-26-2.
- Available in small milligram quantities, e.g., 5 MG.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.