Learn More
Medchemexpress LLC Benzyl N6-(t-Boc)-L-lysinate | 133170-57-7 | MFCD11045868 | 97.5% | 372.89 g/mol | C18H29ClN2O4 | 5 G

Supplier: Medchemexpress LLC HYW0887125G
Benzyl N6-(t-Boc)-L-lysinate is a t-Boc protected lysine derivative used as an intermediate or reagent in peptide synthesis and other organic and biochemical research applications.
- t-Boc protected lysine derivative suitable for synthetic chemistry use.
- White to off-white solid; easy to weigh and handle.
- High purity: 97.5% (LCMS, rounded).
- Molecular formula: C18H29ClN2O4; molecular weight: 372.89 g/mol.
- CAS number: 133170-57-7 for unambiguous identification.
- Storage: store dry and cool; in solution store at -80°C (2 years) or -20°C (1 year).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.