Learn More
Medchemexpress LLC Benzyl N6-(t-Boc)-L-lysinate | 133170-57-7 | MFCD11045868 | 97.5% | 372.89 g/mol | C18H29ClN2O4 | 50 G

Supplier: Medchemexpress LLC HYW08871250G
Benzyl N6-(t-Boc)-L-lysinate is a Boc-protected L-lysine derivative supplied for use in organic synthesis and life-science research, commonly used as a protected amino acid building block in peptide synthesis and related synthetic applications.
- Used as a protected amino acid building block in peptide synthesis.
- Boc-protected lysine derivative suitable for organic synthesis.
- Available in solid form with reported purity of 97.49%.
- CAS number 133170-57-7 for chemical identification.
- Store at room temperature, keep dry and cool; in solution: -80°C (2 years), -20°C (1 year).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.