Learn More
Medchemexpress LLC Benzyl nicotinate | 94-44-0 | 99.6% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB155510MM/1ML
Encompass_Preferred
Benzyl nicotinate is a vasodilator that increases skin oxygenation levels by increasing skin blood flow. It crosses the skin, releases nicotinic acid by enzymatic hydrolysis, and promotes skin oxygenation. It can be used for particulate encapsulation, skin penetration, and detection due to its lipophilicity.
  • Increases skin oxygenation levels
  • Increases skin blood flow
  • Releases nicotinic acid by enzymatic hydrolysis
  • Promotes skin oxygenation
  • Used for particulate encapsulation
  • Used for skin penetration
  • Used for detection due to its lipophilicity
  • Affects lipid structure in the stratum corneum
  • Increases lipid fluidity in the stratum corneum
Catalog No. 50-004-11942