Learn More
Medchemexpress LLC Benzyl nicotinate | 94-44-0 | 99.6% | 5 G
SDP
Supplier:  Medchemexpress LLC HYB15555G
Encompass_Preferred
Benzyl nicotinate is a vasodilator that increases skin oxygenation levels by increasing skin blood flow. It crosses the skin, releases nicotinic acid by enzymatic hydrolysis, increases local cutaneous blood flow, and promotes skin oxygenation. It can be used for particulate encapsulation, skin penetration, and detection for its lipophilicity.
  • Affects the lipid structure and increases the lipid fluidity in the stratum corneum.
  • Exhibits good skin oxygenation levels in BALB/c mouse models.
Catalog No. 50-004-11386