Learn More
Medchemexpress LLC Benzyl-PEG1-Tos | 250MG
SDP
Supplier:  Medchemexpress LLC HY114843250MG
Encompass_Preferred
Benzyl-PEG1-Tos is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs[1]
Catalog No. 50-002-29395