Learn More
Medchemexpress LLC Benzyl-PEG2-CH2COOH | 91842-53-4 | 254.28 | 1 G
SDP
Supplier:  Medchemexpress LLC 1GHY135339
Encompass_Preferred
Benzyl-PEG2-CH2COOH is a PEG-based PROTAC linker designed for the synthesis of PROTACs. PROTACs are chemical compounds that exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. They consist of two different ligands connected by a linker: one for an E3 ubiquitin ligase and another for the target protein.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Enables targeted protein degradation via the ubiquitin-proteasome system
  • For research use only
Catalog No. 50-003-65182