Learn More
Medchemexpress LLC Benzyl-PEG45-alcohol | 26403-74-7 | 98.6% | 2090.50 g/mol | C97H188O46 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY15315310MG
Encompass_Preferred
Benzyl-PEG45-alcohol is a polyethylene glycol (PEG)-based linker bearing a benzyl-protected terminal alcohol, designed for use in the synthesis of proteolysis targeting chimeras (PROTACs). It provides a flexible spacer between ligands and is supplied as a high-purity solid suitable for research applications.
  • PEG-based linker with benzyl-protected terminal alcohol.
  • Designed for use in PROTAC synthesis and bioconjugation.
  • High purity (98.6%) for research-grade applications.
  • Molecular weight approximately 2090.5 g/mol.
  • Supplied as a solid in multiple package sizes for flexible use.
  • Store sealed, away from moisture; follow cold storage guidance for solutions.
Catalog No. 50-002-86037