Learn More
Medchemexpress LLC Benzyl-PEG45-alcohol | 26403-74-7 | C97H188O46 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY153153
Encompass_Preferred
Benzyl-PEG45-alcohol is a PEG-based PROTAC linker essential for the synthesis of PROTACs. These compounds exploit the intracellular ubiquitin-proteasome system for selective degradation of target proteins, achieved by connecting two ligands-one for an E3 ubiquitin ligase and another for the target protein-via this linker.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Designed for selective degradation of target proteins
  • Connects two different ligands: one for an E3 ubiquitin ligase, one for a target protein
Catalog No. 50-003-75358