Learn More
Medchemexpress LLC Benzyl-PEG45-alcohol | 26403-74-7 | C97H188O46 | 5 MG

Supplier: Medchemexpress LLC HY1531535MG
Benzyl-PEG45-alcohol is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
- Purity: 98.56%
- Appearance: Solid
- Color: White to off-white
- For research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.